N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: Boc-Met-OSu, N-Boc-L-methionine N-Succinimidyl Ester

B3433-5G
TCIAB3433-5GEA 970 NOK
TCIAB3433-5G
N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester ≥97.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester
Formel: C₁₄H₂₂N₂O₆S
MW: 346.40 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00042760
CAS-nummer: 3845-64-5
EINECS: 223-341-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 124.0 to 128.0 deg-C
Specific rotation [a]20/D -22.0 to -24.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR