5-Bromo-o-toluidine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

B3687-25G B3687-5G
TCIAB3687-25GEA 1500 NOK
TCIAB3687-25G TCIAB3687-5G
5-Bromo-o-toluidine ≥97.0% (by GC, titration analysis)
5-Bromo-o-toluidine
Formel: C₇H₈BrN
Kokepunkt: 139°C
Smeltepunkt: 33 °C
Tetthet: 1,49 g/cm³ (25 °C)
MDL nummer: MFCD00800678
CAS-nummer: 39478-78-9
EINECS: 254-467-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14e986fd6ef2267efb51b5a6e336a82a0879100f&vid=14e986fd6ef2267efb51b5a6e336a82a0879100f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR