2-Picolyl bromide hydrobromide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

B3772-5G
TCIAB3772-5GEA 915 NOK
TCIAB3772-5G
2-Picolyl bromide hydrobromide ≥98.0% (by HPLC, titration analysis)
2-Picolyl bromide hydrobromide
Formel: C₆H₇Br₂N
MDL nummer: MFCD01863544
CAS-nummer: 31106-82-8
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR