2,2'-Bipiperidine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 2-(Piperidin-2-yl)piperidine

B3982-5G
TCIAB3982-5GEA 3950 NOK
TCIAB3982-5G
2,2'-Bipiperidine ≥98.0% (by titrimetric analysis)
2,2'-Bipiperidine
Formel: C₁₀H₂₀N₂
MW: 168.28 g/mol
Kokepunkt: 93°C
MDL nummer: MFCD01572939
CAS-nummer: 531-67-9
EINECS: 208-514-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR