4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde ≥98.0% (by total nitrogen basis)

Leverandør: TCI

Synonymer: 4,4'-Dibromo-4''-formyltriphenylamine, 4-(Bis(4-bromophenyl)amino)benzaldehyde

B3991-200MG
TCIAB3991-200MGEA 1650 NOK
TCIAB3991-200MG
4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde ≥98.0% (by total nitrogen basis)
4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde
Formel: C₁₉H₁₃Br₂NO
MW: 431,13 g/mol
MDL nummer: MFCD12547118
CAS-nummer: 25069-38-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR