Bis(diisopropylamino)chlorophosphine ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Bis(di-i-propylamino)chlorophosphine

B4035-5G
TCIAB4035-5GEA 2850 NOK
TCIAB4035-5G
Bis(diisopropylamino)chlorophosphine ≥97.0% (by titrimetric analysis)
Bis(diisopropylamino)chlorophosphine
Formel: C₁₂H₂₈ClN₂P
MW: 266,79 g/mol
Kokepunkt: 133°C
Smeltepunkt: 99°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00061482
CAS-nummer: 56183-63-2
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR