3'-Bromo-2,2':5',2''-terthiophene ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 3-Bromo-2,5-di(2-thienyl)thiophene

B4050-1G B4050-5G
TCIAB4050-1GEA 4650 NOK
TCIAB4050-1G TCIAB4050-5G
3'-Bromo-2,2':5',2''-terthiophene ≥97.0% (by GC)
3'-Bromo-2,2':5',2''-terthiophene
Formel: C₁₂H₇BrS₃
CAS-nummer: 105125-00-6

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Amber to Dark green powder to lump to clear liquid
Purity (GC) min. 97.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR