[[(4-tert-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-tert-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: Pyrromethene 597

B4400-200MG
TCIAB4400-200MGEA 2050 NOK
TCIAB4400-200MG
[[(4-tert-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-tert-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥98.0% (by HPLC, total nitrogen)
[[(4-tert-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-tert-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane)
Formel: C₂₂H₃₃BF₂N₂
MW: 374,33 g/mol
MDL nummer: MFCD09753047
CAS-nummer: 137829-79-9

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 526.0 to 531.0 nm (CHCl3)
Absorbance (E1%1cm) min. 1900 (CHCl3, 526.0 to 531.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR