(E)-1,2-Bis(1-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥70% (by HPLC)

Leverandør: TCI

Synonymer: (Eind)(1-Naph)Si=Si(1-Naph)(Eind)

B4421-10MG
TCIAB4421-10MGEA 2175 NOK
TCIAB4421-10MG
(E)-1,2-Bis(1-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥70% (by HPLC)
(E)-1,2-Bis(1-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene
Formel: C₇₆H₁₀₄Si₂
MW: 1073,84 g/mol
CAS-nummer: 1620487-87-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 70.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR