(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)

Leverandør: TCI

Synonymer: (Eind)(2-Naph)Si=Si(2-Naph)(Eind)

B4422-50MG
TCIAB4422-50MGEA 3650 NOK
TCIAB4422-50MG
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene
Formel: C₇₆H₁₀₄Si₂
MW: 1073,84 g/mol
Smeltepunkt: 285°C
CAS-nummer: 1253900-41-2

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR