[[(4-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥97.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: Pyrromethene 580

B4489-200MG
TCIAB4489-200MGEA 2925 NOK
TCIAB4489-200MG
[[(4-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥97.0% (by HPLC)
[[(4-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane)
Formel: C₂₂H₃₃BF₂N₂
MW: 374,33 g/mol
CAS-nummer: 151486-56-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 7.00 to 8.00 %
Melting point 184.0 to 188.0 deg-C
Lambda max. 514.0 to 519.0 nm (CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR