1,3-Bis[(di-tert-butylphosphino)oxy]benzene ≥94.0% (by GC)

Leverandør: TCI
Warning

B4595-1G
TCIAB4595-1GEA 1650 NOK
TCIAB4595-1G
1,3-Bis[(di-tert-butylphosphino)oxy]benzene ≥94.0% (by GC)
1,3-Bis[(di-tert-butylphosphino)oxy]benzene
Formel: C₂₂H₄₀O₂P₂
MW: 398,51 g/mol
CAS-nummer: 338800-20-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR