Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) ≥98.0% (by titrimetric analysis)

Leverandør: TCI

B4837-500MG
TCIAB4837-500MGEA 7850 NOK
TCIAB4837-500MG
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) ≥98.0% (by titrimetric analysis)
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0)
Formel: C₃₂H₅₆N₂P₂Pd
MW: 637,18 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD15071402
CAS-nummer: 1233717-68-4

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystaline
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR