N-Boc-D-prolinal ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate

B5395-1G
TCIAB5395-1GEA 905 NOK
TCIAB5395-1G
N-Boc-D-prolinal ≥95.0% (by GC)
N-Boc-D-prolinal
Formel: C₁₀H₁₇NO₃
Lagringstemperatur: Fryseskap
MDL nummer: MFCD01321389
CAS-nummer: 73365-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dda5ef83daca6b4334397d01663f42fb901e3e4&vid=ce2484f25aaa6cd2a3e9d48e63ab7252988192db&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D +81.0 to +86.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR