Biotin-PEG4-amine ≥94.0% (by HPLC)

Leverandør: TCI

B5560-10MG B5560-50MG
TCIAB5560-10MGEA 620 NOK
TCIAB5560-10MG TCIAB5560-50MG
Biotin-PEG4-amine ≥94.0% (by HPLC)
Biotin-PEG4-amine
Formel: C₂₀H₃₈N₄O₆S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD26142992
CAS-nummer: 663171-32-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 94.0 area%
Elemental analysis (Nitrogen) 11.30 to 13.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR