Biotin-PEG6-Maleimide ≥96.0% (by HPLC)

Leverandør: TCI

B5563-10MG B5563-50MG
TCIAB5563-10MGEA 1650 NOK
TCIAB5563-10MG TCIAB5563-50MG
Biotin-PEG6-Maleimide ≥96.0% (by HPLC)
Biotin-PEG6-Maleimide
Formel: C₃₁H₅₁N₅O₁₁S
Lagringstemperatur: Fryseskap
CAS-nummer: 1808990-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e00182b3c67af4432cec5efde7af2403a6c53ba9&vid=e00182b3c67af4432cec5efde7af2403a6c53ba9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Elemental analysis (Nitrogen) 9.50 to 10.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR