Biotin-PEG11-Amine ≥93.0% (by HPLC)

Leverandør: TCI

B5565-10MG B5565-50MG
TCIAB5565-10MGEA 785 NOK
TCIAB5565-10MG TCIAB5565-50MG
Biotin-PEG11-Amine ≥93.0% (by HPLC)
Biotin-PEG11-Amine
Formel: C₃₄H₆₆N₄O₁₃S
Lagringstemperatur: Fryseskap
CAS-nummer: 1418022-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c48750e2c6a23fce1e97f4aa008973cbacc0d36&vid=2c48750e2c6a23fce1e97f4aa008973cbacc0d36&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Elemental analysis (Nitrogen) 6.50 to 8.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR