Biotin-PEG4-hydrazide ≥94.0% (by HPLC)

Leverandør: TCI

Synonymer: N-[2-[2-[2-[2-(3-Hydrazinyl-3-oxopropoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide, 15-(Biotinamido)-4,7,10,13-tetraoxapentadecanehydrazide, Hydrazide-PEG4-Biotin

B5578-10MG B5578-50MG
TCIAB5578-10MGEA 850 NOK
TCIAB5578-10MG TCIAB5578-50MG
Biotin-PEG4-hydrazide ≥94.0% (by HPLC)
Biotin-PEG4-hydrazide
Formel: C₂₁H₃₉N₅O₇S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD09952658
CAS-nummer: 756525-97-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 94.0 area%
Elemental analysis (Nitrogen) 13.00 to 14.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR