2-Bromo-13,13-dimethyl-6,11-di(naphthalen-2-yl)-13H-indeno[1,2-b]anthracene ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

B5581-1G
TCIAB5581-1GEA 1800 NOK
TCIAB5581-1G
2-Bromo-13,13-dimethyl-6,11-di(naphthalen-2-yl)-13H-indeno[1,2-b]anthracene ≥98.0% (by HPLC, titration analysis)
2-Bromo-13,13-dimethyl-6,11-di(naphthalen-2-yl)-13H-indeno[1,2-b]anthracene
Formel: C₄₃H₂₉Br
MW: 625,61 g/mol
MDL nummer: MFCD28048411
CAS-nummer: 1196107-75-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR