Biotin-HPDP

Leverandør: TCI

B5749-25MG B5749-100MG
TCIAB5749-25MGEA 2500 NOK
TCIAB5749-25MG TCIAB5749-100MG
Biotin-HPDP
Biotin-HPDP
Formel: C₂₄H₃₇N₅O₃S₃
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00078541
CAS-nummer: 129179-83-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 12.30 to 13.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR