SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Leverandør: TCI
B5834-25ML
TCIAB5834-25MLEA 730 NOK
TCIAB5834-25ML
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Buffere Buffere, elektroforese Buffere, ladning
SDS-PAGE Sample Buffer is specially formulated for protein sample preparation to based on the Laemmli Method of SDS-PAGE system.
This product contains 4% SDS, 20% glycerol, 0,01% bromophenol blue and 0,125 M Tris HCl, pH approx. 6,8. Does not contain 2-Mercaptoethanol.
NB: Please use after bringing back to normal temperature.