3-(tert-Butyldimethylsilyloxy)propan-1-amine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-((tert-Butyldimethylsilyl)oxy)propan-1-amine

B5959-1ML
TCIAB5959-1MLEA 2200 NOK
TCIAB5959-1ML
3-(tert-Butyldimethylsilyloxy)propan-1-amine ≥98.0% (by GC, titration analysis)
3-(tert-Butyldimethylsilyloxy)propan-1-amine
Formel: C₉H₂₃NOSi
MW: 189,37 g/mol
Kokepunkt: 70°C
MDL nummer: MFCD18206137
CAS-nummer: 115306-75-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR