Biotin-LC-LC-NHS

Leverandør: TCI

Synonymer: N-Succinimidyl N-[6-(Biotinamido)hexanoyl]-6-aminohexanoate, Biotin-LC-LC-NHS (2mg*5), 2,5-Dioxopyrrolidin-1-yl 6-(6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanamido)hexanoate, N-[6-(Biotinamido)hexanoyl]-6-aminohexanoic acid N-Succinimidyl ester

B6096-1SET
TCIAB6096-1SETEA 1575 NOK
TCIAB6096-1SET
Biotin-LC-LC-NHS
Biotin-LC-LC-NHS
Formel: C₂₆H₄₁N₅O₇S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00467154
CAS-nummer: 89889-52-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Functionality test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR