Biotin-PEG2-NHS

Leverandør: TCI

B6097-1SET
TCIAB6097-1SETEA 1500 NOK
TCIAB6097-1SET
Biotin-PEG2-NHS
Biotin-PEG2-NHS
Formel: C₂₁H₃₂N₄O₈S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00467154
CAS-nummer: 596820-83-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Functionality test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR