3'-Chloroindophenol sodium salt redox indicator

Leverandør: TCI
Warning

C0188-1G
TCIAC0188-1GEA 1325 NOK
TCIAC0188-1G
3'-Chloroindophenol sodium salt redox indicator
3'-Chloroindophenol sodium salt
Formel: C₁₂H₈ClNO₂
MDL nummer: MFCD00059876
CAS-nummer: 41350-02-1

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green to Dark blue to Black powder to crystal
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR