Chrome pure blue BX

Leverandør: TCI

Synonymer: Mordant Blue 1, Eriochrome Azurol B, Sunchromine Pure Blue BX

C0337-25G
TCIAC0337-25GEA 540 NOK
TCIAC0337-25G
Chrome pure blue BX
Chrome pure blue BX
Formel: C₂₃H₁₆Cl₂O₆
MW: 503.24 g/mol
MDL nummer: MFCD00036446
CAS-nummer: 1796-92-5
EINECS: 217-279-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd79256d29eff7e2be5950cd38300f6d3795c410&vid=b7f0f9dab2c050842378461b1d0d8e44b8506270&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Lambda max. 423.0 to 429.0 nm (H2O)
Absorbance (E1%1cm) min. 220 (H2O, 423.0 to 429.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR