cis-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

C0458-25G C0458-500G
TCIAC0458-25GEA 220 NOK
TCIAC0458-25G TCIAC0458-500G
cis-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
cis-1,2-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
Kokepunkt: 339 °C (1013 hPa)
Smeltepunkt: 188…192 °C
MDL nummer: MFCD00064949
CAS-nummer: 610-09-3
EINECS: 445-870-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26ca43447b77710d63a6cc64bcac1c6e8eb57be4&vid=26ca43447b77710d63a6cc64bcac1c6e8eb57be4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 187.0 to 198.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR