L-Cystine dihydrochloride ≥98.0% (by HPLC)

Leverandør: TCI

C0520-25G
TCIAC0520-25GEA 650 NOK
TCIAC0520-25G
L-Cystine dihydrochloride ≥98.0% (by HPLC)
L-Cystine dihydrochloride
Formel: C₆H₁₂N₂O₄S₂·2HCl
MDL nummer: MFCD00070399
CAS-nummer: 30925-07-6
EINECS: 250-391-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -163 to -173 deg (C=1, 1mol/L HCl)
Solubility in 1mol/L HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR