Cytidine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

C0522-1G C0522-25G C0522-5G
TCIAC0522-1GEA 220 NOK
TCIAC0522-1G TCIAC0522-25G TCIAC0522-5G
Cytidine ≥98.0% (by HPLC, titration analysis)
Cytidine
Formel: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006545
CAS-nummer: 65-46-3
EINECS: 200-610-9
Merck Indeks: 14,02786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a464f934275311983fe32edac4443b279f8db7a6&vid=a464f934275311983fe32edac4443b279f8db7a6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +32.0 to +36.0 deg (C=0.7, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR