Cytidine-5'-monophosphate disodium salt hydrate ≥99.0% (by HPLC)

Leverandør: TCI

Synonymer: 5'-Cytidylic Acid Disodium Salt, 5'-CMP 2Na

C0524-1G C0524-5G
TCIAC0524-1GEA 220 NOK
TCIAC0524-1G TCIAC0524-5G
Cytidine-5'-monophosphate disodium salt hydrate ≥99.0% (by HPLC)
Cytidine-5'-monophosphate disodium salt hydrate
Formel: C₉H₁₄N₃O₈P
MW: 367.16 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00064355
CAS-nummer: 6757-06-8
EINECS: 229-819-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=386ddd47e38e0e44fd839ed2e6526271f614eab9&vid=583565fe49920593a19c43b0b172812538feb049&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Specific rotation [a]20/D +7.0 to +9.0 deg (C=2, H2O) (calcd.on anh.substance)
abs-ratio (a/b) 0.40 to 0.50 (E250/E260,0.01mol/L HCl)
abs-ratio (2a/2b) 1.90 to 2.20 (E280/E260,0.01mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR