Eucalyptol ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane

C0542-25ML
TCIAC0542-25MLEA 1975 NOK
TCIAC0542-25ML
Eucalyptol ≥99.0% (by GC)
Eucalyptol
Formel: C₁₀H₁₈O
Kokepunkt: 176°C
Tetthet: 0,92 g/cm³ (20 °C)
Flash punkt: 48°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167977
CAS-nummer: 470-82-6
EINECS: 207-431-5
FN: 1993
ADR: 3,III
Merck Indeks: 14,03895
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41149715b83e603fd6ea3a8ffc18a3c254c18939&vid=41149715b83e603fd6ea3a8ffc18a3c254c18939&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Freezing point min. 1.4 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR