Choline hydrogen tartrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

C0553-25G C0553-500G
TCIAC0553-25GEA 220 NOK
TCIAC0553-25G TCIAC0553-500G
Choline hydrogen tartrate ≥98.0% (by titrimetric analysis)
Choline hydrogen tartrate
Formel: C₉H₁₉NO₇
MDL nummer: MFCD00036332
CAS-nummer: 87-67-2
EINECS: 201-763-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcf4f82c5f57690ad194f7eecf6fb79fac634fad&vid=fcf4f82c5f57690ad194f7eecf6fb79fac634fad&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +16.5 to +17.5 deg (C=10, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR