4-Chlorobenzyl alcohol ≥99.0% (by GC)

Leverandør: TCI

Synonymer: (4-Chlorophenyl)methanol

C0565-25G
TCIAC0565-25GEA 930 NOK
TCIAC0565-25G
4-Chlorobenzyl alcohol ≥99.0% (by GC)
4-Chlorobenzyl alcohol
Formel: C₇H₇ClO
MW: 142.58 g/mol
Kokepunkt: 119°C
MDL nummer: MFCD00004652
CAS-nummer: 873-76-7
EINECS: 212-852-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=027fa1c2017553cdd13d592a77d0df3c831f3511&vid=027fa1c2017553cdd13d592a77d0df3c831f3511&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 71.0 to 73.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR