2-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)

Leverandør: TCI

C0754-25G
TCIAC0754-25GEA 905 NOK
TCIAC0754-25G
2-Chlorophenylhydrazine sulphate ≥95.0% (by titrimetric analysis)
2-Chlorophenylhydrazine sulphate
Formel: C₁₂H₁₄Cl₂N₄.H₂SO₄
MDL nummer: MFCD00060255
CAS-nummer: 198279-94-6

Bestill nå

Spesifikasjon Testresultater

Purity (Potassium iodate Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR