cis-1,4-Cyclohexanedicarboxylic acid ≥90.0% (by GC)

Leverandør: TCI
Warning

Synonymer: cis-Cyclohexane-1,4-dicarboxylic acid

C0789-100G C0789-25G
TCIAC0789-100GEA 6200 NOK
TCIAC0789-100G TCIAC0789-25G
cis-1,4-Cyclohexanedicarboxylic acid ≥90.0% (by GC)
cis-1,4-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
MDL nummer: MFCD00064950
CAS-nummer: 619-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e3a1fa79c7df8cf063265920825c510af7ff1ff&vid=5e3a1fa79c7df8cf063265920825c510af7ff1ff&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 167.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR