trans-2-Hexenyl hexanoate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: Hexanoic Acid trans-2-Hexenyl Ester

C0904-25ML
TCIAC0904-25MLEA 1025 NOK
TCIAC0904-25ML
trans-2-Hexenyl hexanoate ≥97.0% (by GC)
trans-2-Hexenyl hexanoate
Formel: C₁₂H₂₂O₂
MW: 198,31 g/mol
Kokepunkt: 79°C
Flash punkt: 104°C
MDL nummer: MFCD00036551
CAS-nummer: 53398-86-0
EINECS: 258-519-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR