2-Chloro-5-nitrobenzotrifluoride ≥97.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Chloro-5-nitrobenzotrifluoride, 2-Chloro-α,α,α-trifluoro-5-nitrotoluene, 1-Chloro-4-nitro-2-(trifluoromethyl)benzene

C0923-25G
TCIAC0923-25GEA 270 NOK
TCIAC0923-25G
2-Chloro-5-nitrobenzotrifluoride ≥97.0% (by GC)
2-Chloro-5-nitrobenzotrifluoride
Formel: C₇H₃ClF₃NO₂
MW: 225,55 g/mol
Kokepunkt: 232 °C (1013 hPa)
Smeltepunkt: 25…27 °C
Tetthet: 1,527 g/cm³ (20 °C)
MDL nummer: MFCD00007296
CAS-nummer: 777-37-7
EINECS: 212-287-1
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68a9397fa49b28dee52a8b582f0519c4af953717&vid=68a9397fa49b28dee52a8b582f0519c4af953717&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Freezing point 20.0 to 23.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR