3,3-Diethoxypropiononitrile ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3,3-Diethoxypropionitrile, 3,3-Diethoxypropanenitrile, Cyanoacetaldehyde diethyl acetal

C1178-10ML
TCIAC1178-10MLEA 1025 NOK
TCIAC1178-10ML
3,3-Diethoxypropiononitrile ≥95.0% (by GC)
3,3-Diethoxypropiononitrile
Formel: C₇H₁₃NO₂
MW: 143.19 g/mol
Kokepunkt: 99°C
Flash punkt: 84°C
MDL nummer: MFCD00040558
CAS-nummer: 2032-34-0
EINECS: 217-988-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96c385b3af5ed8e30acf66517c57360b0919e60f&vid=96c385b3af5ed8e30acf66517c57360b0919e60f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR