4-Chloroindole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Chloro-1H-indole

C1248-1G
TCIAC1248-1GEA 835 NOK
TCIAC1248-1G
4-Chloroindole ≥98.0% (by GC)
4-Chloroindole
Formel: C₈H₆ClN
MW: 151,6 g/mol
Kokepunkt: 150°C
Smeltepunkt: 84…86 °C
Tetthet: 1,259 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005665
CAS-nummer: 25235-85-2
EINECS: 246-747-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e306703fb8765fe031646b145543478ed283af13&vid=e306703fb8765fe031646b145543478ed283af13&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR