Cyclopentanone oxime ≥98.0% (by GC)

Leverandør: TCI

C1307-25G
TCIAC1307-25GEA 835 NOK
TCIAC1307-25G
Cyclopentanone oxime ≥98.0% (by GC)
Cyclopentanone oxime
Formel: C₅H₉NO
Kokepunkt: 97°C
MDL nummer: MFCD00001420
CAS-nummer: 1192-28-5
EINECS: 214-749-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ae6731900e5db798a2a7119c8bd13f16d3e78e2&vid=6ae6731900e5db798a2a7119c8bd13f16d3e78e2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR