4-kolesten-3-on ≥95.0% (by GC)

Leverandør: TCI

Synonymer: (8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one, (+)-4-Cholesten-3-one

C1366-5G
TCIAC1366-5GEA 1525 NOK
TCIAC1366-5G
4-kolesten-3-on ≥95.0% (by GC)
4-kolesten-3-on
Formel: C₂₇H₄₄O
MW: 384,65 g/mol
MDL nummer: MFCD00003663
CAS-nummer: 601-57-0
EINECS: 210-005-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 79.0 to 83.0 deg-C
Specific rotation [a]20/D +91.0 to +95.0 deg (C=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR