(S)-(+)-Camptothecin 97,0 HPLC_ASSAY_METHOD

Leverandør: TCI
Danger

Synonymer: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

C1495-1G C1495-100MG
TCIAC1495-1GEA 2675 NOK
TCIAC1495-1G TCIAC1495-100MG
(S)-(+)-Camptothecin 97,0 HPLC_ASSAY_METHOD
(S)-(+)-Camptothecin
Formel: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
MDL nummer: MFCD00081076
CAS-nummer: 7689-03-4
FN: 1544
ADR: 6.1 III
Merck Indeks: 14,01735

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +37.0 to +42.0 deg (C=0.4, CHCl3:MeOHl=8:2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR