1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecane, 1,4-Cyclohexanedione bis(ethylene acetal), 1,4-Cyclohexanedione bisethylene ketal

C1544-25G
TCIAC1544-25GEA 1650 NOK
TCIAC1544-25G
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0% (by GC)
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan
Formel: C₁₀H₁₆O₄
MW: 200,23 g/mol
MDL nummer: MFCD00010851
CAS-nummer: 183-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b478cbfb7baefc61b3870761155efeaa47f00af&vid=6b478cbfb7baefc61b3870761155efeaa47f00af&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 77.0 to 81.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR