(1-Ethoxycarbonylethylidene)triphenylphosphorane ≥97.0% (by HPLC)

Leverandør: TCI

C1641-5G
TCIAC1641-5GEA 1075 NOK
TCIAC1641-5G
(1-Ethoxycarbonylethylidene)triphenylphosphorane ≥97.0% (by HPLC)
(1-Ethoxycarbonylethylidene)triphenylphosphorane
Formel: C₂₃H₂₃O₂P
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00009160
CAS-nummer: 5717-37-3
EINECS: 227-215-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c355de49a23571d275edb99e579f90840414b26&vid=6c355de49a23571d275edb99e579f90840414b26&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR