anti-(1R)-(+)-Camphorquinone 3-oxime ≥95.0% (by GC, total nitrogen)

Leverandør: TCI

Synonymer: anti-(1R)-(+)-Camphorquinone 3-Oxime, (1R,E)-(+)-2,3-Bornanedione 3-oxime,anti-(1R)-(+)-Camphorquinone 3-oxime, anti-(1R)-(+)-2,3-Bornanedione 3-Oxime

C1661-1G
TCIAC1661-1GEA 1175 NOK
TCIAC1661-1G
anti-(1R)-(+)-Camphorquinone 3-oxime ≥95.0% (by GC, total nitrogen)
anti-(1R)-(+)-Camphorquinone 3-oxime
Formel: C₁₀H₁₅NO₂
MW: 181.24 g/mol
MDL nummer: MFCD00074848
CAS-nummer: 31571-14-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (with Total Nitrogen) min. 95.0 %
Melting point 153.0 to 157.0 deg-C
Specific rotation [a]20/D +195 to +201 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR