Cytidine-5'-monophosphate ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 5'-Cytidylic Acid, 5'-CMP, ((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

C1675-5G C1675-1G
TCIAC1675-5GEA 1200 NOK
TCIAC1675-5G TCIAC1675-1G
Cytidine-5'-monophosphate ≥98.0% (by HPLC)
Cytidine-5'-monophosphate
Formel: C₉H₁₄N₃O₈P
MW: 323.20 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006544
CAS-nummer: 63-37-6
EINECS: 200-556-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da9ef97850d86effd034326d49b2202314bbddc9&vid=16aa6809a45d7d58e713ccf89f1475afee00134e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Specific rotation [a]20/D +9.0 to +10.5 deg (C=1, 0.5mol/L Na2HPO4 sol.)
abs-ratio (a/b) 0.40 to 0.50 (E250/E260)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR