N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

C1767-500MG
TCIAC1767-500MGEA 2050 NOK
TCIAC1767-500MG
N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)
N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d4b493dab785522ce4ddacbd08b649985c0dbfd&vid=354239786257edc89f3179368a6850346c7bcaa0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +90.0 to +100.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR