(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: (1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid

C1953-1G C1953-5G
TCIAC1953-1GEA 675 NOK
TCIAC1953-1G TCIAC1953-5G
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
MW: 172.18 g/mol
MDL nummer: MFCD00062973
CAS-nummer: 46022-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e0149b729a851f6abffd4b3b0a3a4f867504341&vid=2e0149b729a851f6abffd4b3b0a3a4f867504341&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (GC) min. 95.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 185.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR