1-(2-Cyanoethyl)-2-phenylimidazole ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-(2-Phenyl-1-imidazolyl)propionitrile

C1964-25G
TCIAC1964-25GEA 475 NOK
TCIAC1964-25G
1-(2-Cyanoethyl)-2-phenylimidazole ≥98.0% (by GC, titration analysis)
1-(2-Cyanoethyl)-2-phenylimidazole
Formel: C₁₂H₁₁N₃
MW: 197,24 g/mol
CAS-nummer: 23996-12-5
EINECS: 245-972-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR