Carboplatin ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)

C2043-1G C2043-100MG
TCIAC2043-1GEA 4200 NOK
TCIAC2043-1G TCIAC2043-100MG
Carboplatin ≥98.0% (by HPLC)
Carboplatin
Formel: C₆H₁₂N₂O₄Pt
MW: 371.26 g/mol
MDL nummer: MFCD00070464
CAS-nummer: 41575-94-4
EINECS: 255-446-0
Merck Indeks: 14,01822

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR